Potassium trioxalatoferrate(III) trihydrate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
31124.14
31124.22
31124.06
31124.36
31124.14EA
3070
CZK
31124.14
31124.22
31124.06
31124.36
Potassium trioxalatoferrate(III) trihydrate
Potassium trioxalatoferrate(III) trihydrate
CAS No.: 5936-11-8
Molecular Formula: K3Fe(C2O4)3.3H2O
Formula Weight: 491.26 (437.21anhy)
Physical Form: Crystalline
Density: 2.133
Sensitivity: Hygroscopic
Appearance: Green
UN No.: UN3288
MDL No.: MFCD00150450
Vzorec:
K₃Fe(C₂O₄)₃·3H₂O Hustota: 2,133 Storage Temperature: Ambient |
MDL Number:
MFCD00150450 CAS číslo: 5936-11-8 EINECS: 238-954-7 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...