Tisk…

Carnauba wax

Dodavatel: Thermo Scientific

31117.30 31117.A1 31117.A7
31117.30EA 1560 CZK
31117.30 31117.A1 31117.A7
Carnauba wax
Carnauba wax

CAS No.: 8015-86-9 Physical Form: Flakes Density: 0.990-0.999 Refractive Index: 1.4500 Odour: Odourless Melting Point: 82-86° MDL No.: MFCD00130724

Bod tání: 82…86 °C
Hustota: 0,990…0.999 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00130724
CAS číslo: 8015-86-9
EINECS: 232-399-4
Merck Index: 14,1846

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR