Linear alkylbenzenesulphonic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
30574.30
30574.A1
30574.D5
30574.30EA
1360
CZK
30574.30
30574.A1
30574.D5
Linear alkylbenzenesulphonic acid ≥97%
Linear alkylbenzenesulphonic acid
Purity: 97%
CAS No.: 68584-22-5
Molecular Formula: RC6H4SO3H
Physical Form: Liquid
UN No.: UN2586
MDL No.: MFCD00147445
Vzorec:
C₁₈H₃₀O₃S Storage Temperature: Ambient |
MDL Number:
MFCD00147445 CAS číslo: 68584-22-5 EINECS: 271-528-9 UN: 2586 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...