Iron(II) oxide ≥99.5% (metals basis)
Dodavatel: Thermo Scientific
30513.06
30513.14
30513.06EA
6900
CZK
30513.06
30513.14
Iron(II) oxide ≥99.5% (metals basis)
Iron(II) oxide
Purity: 99.5%
CAS No.: 1345-25-1
Molecular Formula: FeO (may contain trace amounts of other phases)
Formula Weight: 71.85
Physical Form: -10 Mesh
Density: 5.7
Sensitivity: Air Sensitive
Appearance: Black
Odour: Odourless
Melting Point: 1369°
Solubility: Insoluble in water and alkalies. Soluble in HCl, H2SO4. Slightly soluble in HNO3
MDL No.: MFCD00016095
Applications: In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalyst
Vzorec:
FeO MW: 71,844 g/mol Bod tání: 1369 °C Hustota: 5,7 Storage Temperature: Ambient |
MDL Number:
MFCD00016095 CAS číslo: 1345-25-1 EINECS: 215-721-8 Merck Index: 14,4052 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...