Calcium tetrafluoroborate hydrate
Dodavatel: Thermo Scientific
|
Nebezpečí
|
30131.14
30131.22
30131.14EA
6040
CZK
30131.14
30131.22
Calcium tetrafluoroborate hydrate
Calcium tetrafluoroborate hydrate
CAS No.: 15978-68-4
Molecular Formula: Ca(BF4)2.xH2O
Formula Weight: 213.68(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00015972
Vzorec:
Ca(BF₄)₂·xH₂O MW: 213,68 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00015972 CAS číslo: 13814-93-2 EINECS: 237-485-5 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...