Tisk…
Copper(II) tetrafluoroborate hexahydrate ≥98%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
26127.22 26127.36
26127.22EA 5190 CZK
26127.22 26127.36
Copper(II) tetrafluoroborate hexahydrate ≥98%
Copper(II) tetrafluoroborate hexahydrate
Purity: 98% CAS No.: 72259-10-0 Molecular Formula: Cu(BF4)2.6H2O Formula Weight: 345.25 (237.15anhy) Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: UN3260 MDL No.: MFCD00150223
Vzorec: B₂CuF₈H₁₂O₆
MW: 345.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150223
CAS číslo: 72259-10-0
EINECS: 253-959-4
UN: 3260
ADR: 8 III