Tisk…

Strontium dodecairon nonadecaoxide ≥99.5% (metals basis)

Dodavatel: Thermo Fisher Scientific

22569.06 22569.14
22569.06EA 2030 CZK
22569.06 22569.14
Strontium dodecairon nonadecaoxide ≥99.5% (metals basis)
Strontium dodecairon nonadecaoxide

Purity: 99.5% CAS No.: 12023-91-5 Molecular Formula: SrFe12O19 Formula Weight: 1061.77 Physical Form: -325 Mesh Powder Melting Point: 450° MDL No.: MFCD00075632

Vzorec: SrFe₁₂O₁₉
MW: 1061.77 g/mol
Bod tání: >450 °C
Storage Temperature: Ambient
MDL Number: MFCD00075632
CAS číslo: 12023-91-5
EINECS: 234-685-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR