Tisk…

Sodium metatungstate monohydrate

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate

22534.18 22534.30 22534.56
22534.18EA 2800 CZK
22534.18 22534.30 22534.56
Sodium metatungstate monohydrate
Sodium metatungstate monohydrate

CAS No.: 12141-67-2 Molecular Formula: Na6O39W12.H2O Formula Weight: 2986.13 (2968.12anhy) Physical Form: Powder Density: 5.47 Appearance: White Odour: Odourless MDL No.: MFCD00061466

Vzorec: Na₆O₃₉W₁₂·H₂O
MW: 2986.13 g/mol (2968.12 anhy)
Hustota: 5.47
Storage Temperature: Ambient
MDL Number: MFCD00061466
CAS číslo: 12141-67-2
EINECS: 412-770-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR