Tisk…

Zinc ferrite

Dodavatel: Thermo Fisher Scientific

19407.14 19407.22
19407.14EA 3610 CZK
19407.14 19407.22
Zinc ferrite
Zinc ferrite

CAS No.: 12063-19-3 Molecular Formula: ZnFe2O4 Formula Weight: 241.06 Physical Form: Fine Powder Odour: Odourless MDL No.: MFCD00049621

Vzorec: Fe₂H₂O₄Zn
Storage Temperature: Ambient
MDL Number: MFCD00049621
CAS číslo: 12063-19-3
EINECS: 235-052-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR