Zinc trifluoroacetate hydrate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
18686.14
18686.22
18686.14EA
3210
CZK
18686.14
18686.22
Zinc trifluoroacetate hydrate
Zinc trifluoroacetate hydrate
CAS No.: 207801-31-8
Molecular Formula: Zn(OOCCF3)2.xH2O
Formula Weight: 291.40(anhy)
Physical Form: Powder/chunks
Sensitivity: Hygroscopic
UN No.: UN3077
MDL No.: MFCD00061595
Vzorec:
C₄H₂F₆O₅Zn MW: 309.425 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061595 CAS číslo: 207801-31-8 UN: 3077 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...