Tisk…

2,4,6-Triisopropylbenzenesulphonyl hydrazide

Dodavatel: Thermo Scientific
Varování

Synonyms: 2,4,6-Triisopropylbenzenesulphonohydrazide

17976.04 17976.02
17976.04EA 3260 CZK
17976.04 17976.02
2,4,6-Triisopropylbenzenesulphonyl hydrazide
2,4,6-Triisopropylbenzenesulphonyl hydrazide

CAS No.: 39085-59-1 Molecular Formula: C15H26N2O2S Formula Weight: 298.58 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 110-112° dec. Beilstein Reference: 2145001 MDL No.: MFCD00014750


Applications: As a reagent for generation of diimide

Vzorec: C₁₅H₂₆N₂O₂S
MW: 298,45 g/mol
Bod tání: 110…112 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00014750
CAS číslo: 39085-59-1
EINECS: 254-282-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR