Ruthenium(III) nitrosyl chloride (≥42,1% Ru)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Trichloronitrosylruthenium(III), Ruthenium(III) nitrosylchloride
16622.03
16622.06
16622.03EA
5290
CZK
16622.03
16622.06
Ruthenium(III) nitrosyl chloride (≥42,1% Ru)
Ruthenium(III) nitrosyl chloride
CAS No.: 18902-42-6
Molecular Formula: Ru(NO)Cl3
Formula Weight: 237.44
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00049765
Vzorec:
Ru(NO)Cl₃ Storage Temperature: Ambient |
MDL Number:
MFCD00049765 CAS číslo: 32535-76-5 EINECS: 242-651-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...