Tisk…

Trimethylolpropane triacrylate stabilizovaný

Dodavatel: Thermo Scientific
Varování

Synonyms: TMPTA, 1,1,1-Tris(acryloyloxymethyl)propane

16445.22 16445.36 16445.A3
16445.22EA 1650 CZK
16445.22 16445.36 16445.A3
Trimethylolpropane triacrylate stabilizovaný
Trimethylolpropane triacrylate

CAS No.: 15625-89-5 Molecular Formula: (CH2=CHCO2CH2)3CCH2CH3 Formula Weight: 296.31 Flash Point: 87°(188°F) Physical Form: Liquid Density: 1.10 Refractive Index: 1.4735 Odour: Pungent Boiling Point: 200°/1mm MDL No.: MFCD00008628

Vzorec: C₁₅H₂₀O₆
MW: 296,32 g/mol
Bod varu: >200 °C (1 mmHg)
Hustota: 1,10
Bod vzplanutí: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008628
CAS číslo: 15625-89-5
EINECS: 239-701-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR