Neodymium(III) carbonate hydrate ≥99.9%
Dodavatel: Thermo Fisher Scientific
15301.18
15301.30
15301.18EA
5150
CZK
15301.18
15301.30
Neodymium(III) carbonate hydrate ≥99.9%
Neodymium(III) carbonate hydrate
Purity: 99.9%
CAS No.: 38245-38-4
Molecular Formula: Nd2(CO3)3.xH2O
Formula Weight: 468.51(anhy)
Physical Form: Powder
MDL No.: MFCD00150426
Vzorec:
Nd₂(CO₃)₃.xH₂O MW: 468.51 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150426 CAS číslo: 38245-38-4 EINECS: 227-579-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...