Silicon tetraacetate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Silicon(IV) acetate
14703.18
14703.30
14703.18EA
4400
CZK
14703.18
14703.30
Silicon tetraacetate ≥97%
Silicon tetraacetate
CAS No.: 562-90-3
Molecular Formula: Si(OOCCH3)4
Formula Weight: 264.27
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 110°
Boiling Point: 160-170° dec.
UN No.: UN3261
MDL No.: MFCD00026184
Vzorec:
C₈H₁₂O₈Si MW: 264.261 g/mol Bod varu: 160…170 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00026184 CAS číslo: 562-90-3 EINECS: 209-239-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...