Barium acetylacetonate hydrate
Dodavatel: Thermo Scientific
|
Varování
|
14613.14
14613.22
14613.14EA
2880
CZK
14613.14
14613.22
Barium acetylacetonate hydrate
Barium acetylacetonate hydrate
CAS No.: 12084-29-6
Molecular Formula: C10H14BaO4.xH2O
Formula Weight: 335.56(anhy)
Physical Form: Powder
Melting Point:
Vzorec:
C₁₀H₁₄BaO₄·xH₂O MW: 335,545 g/mol (anhydrous) Bod tání: >320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013484 CAS číslo: 12084-29-6 EINECS: 235-151-3 UN: 1564 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...