Yttrium(III) oxalate nonahydrate ≥99.9% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Varování
|
14562.18
14562.30
14562.18EA
4330
CZK
14562.18
14562.30
Yttrium(III) oxalate nonahydrate ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) oxalate nonahydrate
Purity: 99.9%
CAS No.: 13266-82-5
Molecular Formula: C6O12Y2.9H2O
Formula Weight: 604.01 (441.87anhy)
Physical Form: Crystalline
Appearance: White
Melting Point: dec.
UN No.: UN3288
MDL No.: MFCD00150502
Vzorec:
C₆O₁₂Y₂·9H₂O MW: 604,01 g/mol Bod tání: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00150502 CAS číslo: 13266-82-5 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...