Yttrium(III) sulphate octahydrate ≥97% (REO, rare earth oxide basis)
Dodavatel: Thermo Scientific
|
Varování
|
14561.18
14561.30
14561.18EA
2850
CZK
14561.18
14561.30
Yttrium(III) sulphate octahydrate ≥97% (REO, rare earth oxide basis)
Yttrium(III) sulphate octahydrate
Purity: 99.9%
CAS No.: 7446-33-5
Molecular Formula: Y2(SO4)3.8H2O
Formula Weight: 610.11 (465.99anhy)
Physical Form: Crystalline
Appearance: White to pale yellow
Odour: Odourless
Solubility: Water solubility decreases with increasing temperature
MDL No.: MFCD00149946
Vzorec:
H₁₆O₂₀S₃Y₂ MW: 610,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149946 CAS číslo: 7446-33-5 EINECS: 236-844-3 Merck Index: 14,10107 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...