Tisk…

Nickel(II) formate dihydrate

Dodavatel: Thermo Scientific
Nebezpečí

13801.22 13801.36 13801.A3
13801.22EA 1580 CZK
13801.22 13801.36 13801.A3
Nickel(II) formate dihydrate
Nickel(II) formate dihydrate

CAS No.: 15694-70-9 Molecular Formula: Ni(HCO2)2.2H2O Formula Weight: 184.77 (148.74anhy) Physical Form: Powder Density: 2.15 Melting Point: 130-140° -2H2O Solubility: In the range 180-200° decomposes to Ni, CO, CO2, H2, H2O, CH4. Partially soluble in water UN No.: UN3077 MDL No.: MFCD00153034


Applications: Nickel catalyst preparations

Vzorec: Ni(HCO₂)₂·2H₂O
MW: 184,77 g/mol (148,74 anhy)
Bod tání: 130…140 °C (-2H₂O)
Hustota: 2,15
Storage Temperature: Ambient
MDL Number: MFCD00153034
CAS číslo: 15694-70-9
EINECS: 222-101-0
UN: 3077
ADR: 9,III
Merck Index: 14,6509

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR