Tisk…

Diamond (synthetic) ≥99.9% (metals basis), powder 40-60 micron

Dodavatel: Thermo Fisher Scientific

13402.03 13402.06
13402.03EA 4400 CZK
13402.03 13402.06
Diamond (synthetic) ≥99.9% (metals basis), powder 40-60 micron
Diamond

Purity: 99.9% MDL No.: MFCD00211867


Applications: Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

Storage Temperature: Ambient
MDL Number: MFCD00211867
CAS číslo: 7782-40-3
EINECS: 231-953-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR