Tisk…

Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Lanthanum(III) acetylacetonate hydrate, Tris(2,4-pentanedionato)lanthanum(III) hydrate, Acetylacetone Lanthanum(III) hydrate

13194.14 13194.22
13194.14EA 4050 CZK
13194.14 13194.22
Lanthanum(III) 2,4-pentanedionate hydrate ≥99.9%
Lanthanum(III) 2,4-pentanedionate hydrate

Purity: 99.9% CAS No.: 64424-12-0 Molecular Formula: C15H21LaO6.xH2O Formula Weight: 436.24(anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to pale yellow Melting Point: 143° MDL No.: MFCD00149057

Vzorec: C₁₅H₂₁LaO₆·xH₂O
MW: 436.24 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149057
CAS číslo: 64424-12-0
EINECS: 238-187-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR