Lutetium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
13189.03
13189.06
13189.03EA
5940
CZK
13189.03
13189.06
Lutetium(III) 2,4-pentanedionate ≥99.9% (REO, rare earth oxide basis), REacton®
Lutetium(III) 2,4-pentanedionate
Purity: 99.9%
CAS No.: 17966-84-6
Molecular Formula: C15H21LuO6
Formula Weight: 472.30
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00078030
Vzorec:
C₁₅H₂₁LuO₆ MW: 472.30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078030 CAS číslo: 17966-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...