Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
13029.14
13029.22
13029.14EA
5050
CZK
13029.14
13029.22
Bismuth germanium oxide ≥99.9995% (metals basis), Puratronic®
Bismuth germanium oxide
Purity: 99.9995%
CAS No.: 12233-56-6
Molecular Formula: Bi4Ge3O12
Formula Weight: 1245.68
Physical Form: Powder
Appearance: White
Odour: Odourless
MDL No.: MFCD00064629
Vzorec:
Bi₄Ge₃O₁₂ MW: 1245.68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064629 CAS číslo: 12233-56-6 EINECS: 235-457-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...