Zinc orthophosphate ≥99.995% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
Synonyms:
Zincphosphate, Zinc phosphate (ortho)
13013.06
13013.14
13013.06EA
3040
CZK
13013.06
13013.14
Zinc orthophosphate ≥99.995% (metals basis), Puratronic®
Zinc orthophosphate
Purity: 99.995%
CAS No.: 7779-90-0
Molecular Formula: O8P2Zn3
Formula Weight: 386.05
Physical Form: Powder
Density: 3.75
Appearance: White
Odour: Odourless
UN No.: UN3077
MDL No.: MFCD00036282
Vzorec:
O₈P₂Zn₃ MW: 386.08 g/mol Hustota: 3.75 Storage Temperature: Ambient |
MDL Number:
MFCD00036282 CAS číslo: 7779-90-0 EINECS: 231-944-3 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...