Tisk…

Zinc oxalate dihydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Zinc(II) ethane-1,2-dioate dihydrate

12985.14 12985.22
12985.14EA 6040 CZK
12985.14 12985.22
Zinc oxalate dihydrate ≥99.999% (metals basis), Puratronic®
Zinc oxalate dihydrate

Purity: 99.999% CAS No.: 4255-07-6 Molecular Formula: C2O4Zn.2H2O Formula Weight: 189.42 (153.39anhy) Physical Form: Powder Density: 3.28 Appearance: White UN No.: UN3288 MDL No.: MFCD00150611

Vzorec: ZnC₂O₄·2H₂O
MW: 189.42 g/mol (153.39 anhy)
Hustota: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00150611
CAS číslo: 55906-21-3
EINECS: 208-934-2
UN: 3288
ADR: 6.1 III
Merck Index: 14,10146

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR