Bis(dibenzylideneacetone)palladium
Dodavatel: Thermo Fisher Scientific
Synonyms:
Bis(dibenzylideneacetone)palladium(0)
Packaged under argon.
CAS No.: 32005-36-0
Molecular Formula: C34H28O2Pd
Formula Weight: 575.00
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Brown to purple
Melting Point: 135° dec.
Solubility: Slightly soluble in CH2Cl2, CHCl3 and benzene
MDL No.: MFCD00051942
Applications: Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation
Vzorec:
C₃₄H₂₈O₂Pd MW: 575 g/mol Bod tání: 135 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00051942 CAS číslo: 32005-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...