Tisk…

Tris(dibenzylideneacetone)dipalladium(0) (≥21,5% Pd)

Dodavatel: Thermo Fisher Scientific

12760.03 12760.06
12760.03EA 3260 CZK
12760.03 12760.06
Tris(dibenzylideneacetone)dipalladium(0) (≥21,5% Pd)
Tris(dibenzylideneacetone)dipalladium(0)

CAS No.: 51364-51-3 Molecular Formula: C51H42O3Pd2 Formula Weight: 915.74 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Purple Melting Point: 152-155° Solubility: Soluble in chlorinated solvents MDL No.: MFCD00013310

Vzorec: C₅₁H₄₂O₃Pd₂
Bod tání: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00013310
CAS číslo: 51364-51-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR