Tisk…

Iron(II) chloride hydrate ≥99%, Reagent Grade

Dodavatel: Thermo Scientific
Nebezpečí

12484.14 12484.30 12484.A1 12484.A7
12484.14EA 999 CZK
12484.14 12484.30 12484.A1 12484.A7
Iron(II) chloride hydrate ≥99%, Reagent Grade
Iron(II) chloride hydrate

Purity: 99% CAS No.: 23838-02-0 Molecular Formula: Cl2Fe.xH2O Formula Weight: 126.75(anhy) Physical Form: Crystalline Sensitivity: Air Sensitive Appearance: Green-yellow Odour: Odourless Solubility: Soluble in water, alcohol and acetone. Insoluble in ether UN No.: UN3260 MDL No.: MFCD00149711


Applications: Reducing agent in metallurgy, in pharmaceutical preparations, as mordant in dyeing and in sewage treatment

Vzorec: Cl₂Fe·xH₂O
MW: 126,745 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149711
CAS číslo: 23838-02-0
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 14,4043

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR