Tisk…

Chromium nitride

Dodavatel: Thermo Scientific
Varování

12149.14 12149.22
12149.14EA 2170 CZK
12149.14 12149.22
Chromium nitride
Chromium nitride

CAS No.: 12053-27-9 Molecular Formula: Mixture of Cr2N and CrN Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00016048

Vzorec: Mixture of Cr₂N and CrN
Storage Temperature: Ambient
MDL Number: MFCD00016048
CAS číslo: 12053-27-9
EINECS: 235-002-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR