Rhodium(III) bromide hydrate
Dodavatel: Thermo Scientific
|
Varování
|
11816.03
11816.06
11816.03EA
27250
CZK
11816.03
11816.06
Rhodium(III) bromide hydrate
Rhodium(III) bromide hydrate
CAS No.: 123333-87-9
Molecular Formula: RhBr3.xH2O
Formula Weight: 342.62(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00149837
Vzorec:
RhBr₃·xH₂O MW: 342,62 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149837 CAS číslo: 123333-87-9 EINECS: 239-687-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...