Ruthenium(III) iodide (≥20,5% Ru), anhydrous
Dodavatel: Thermo Fisher Scientific
|
Varování
|
11805.03
11805.06
11805.03EA
2780
CZK
11805.03
11805.06
Ruthenium(III) iodide (≥20,5% Ru), anhydrous
Ruthenium(III) iodide
CAS No.: 13896-65-6
Molecular Formula: RuI3
Formula Weight: 481.78
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: 590° dec.
MDL No.: MFCD00016316
Vzorec:
I₃Ru Storage Temperature: Ambient |
MDL Number:
MFCD00016316 CAS číslo: 13896-65-6 EINECS: 237-664-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...