Silver phosphate ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
11415.14
11415.06
11415.14EA
3610
CZK
11415.14
11415.06
Silver phosphate ≥99%
Silver phosphate
Purity: 99%
CAS No.: 7784-09-0
Molecular Formula: Ag3O4P
Formula Weight: 418.58
Physical Form: Powder
Density: 6.37
Sensitivity: Light Sensitive
Appearance: Yellow
Odour: Odourless
Melting Point: 849°
Solubility: Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3, ammonia, ammonium carbonate, alkali cyanides and thiosulfates
UN No.: UN3077
MDL No.: MFCD00014148
Applications: In photography
Vzorec:
Ag₃O₄P MW: 418,58 g/mol Bod tání: 849 °C Hustota: 6,37 Storage Temperature: Ambient |
MDL Number:
MFCD00014148 CAS číslo: 7784-09-0 EINECS: 232-049-0 ADR: 9,III Merck Index: 14,8525 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...