Tisk…

Holmium(III) chloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Holmium trichloride

11276.09 11276.18
11276.09EA 6490 CZK
11276.09 11276.18
Holmium(III) chloride, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Holmium(III) chloride

Purity: 99.99% Molecular Formula: HoCl3.xH2O Formula Weight: 271.29(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Melting Point: 718° MDL No.: MFCD00011051

Vzorec: HoCl₃·xH₂O
MW: 271.29
Bod varu: 1500
Bod tání: 718 °C
Hustota: 3.7
Storage Temperature: Ambient
MDL Number: MFCD00011051
CAS číslo: 10138-62-2
EINECS: 233-387-1
Merck Index: 14,4727

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR