Tisk…

Gallium(III) sulphate hydrate ≥99.999% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific

11152.09 11152.18
11152.09EA 6940 CZK
11152.09 11152.18
Gallium(III) sulphate hydrate ≥99.999% (metals basis), Puratronic®
Gallium(III) sulphate hydrate

Purity: 99.999% CAS No.: 13780-42-2 Molecular Formula: Ga2(SO4)3.xH2O (x≈18) Formula Weight: 427.62(anhy) Physical Form: Crystalline Aggregates Density: 3.860 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 105-110° MDL No.: MFCD00149725

Vzorec: Ga₂(SO₄)₃·xH₂O (x≈18)
Bod tání: 105…110 °C
Hustota: 3,860
Storage Temperature: Ambient
MDL Number: MFCD00149725
CAS číslo: 13780-42-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR