Tetraammineplatinum(II) chloride monohydrate ≥98% (metals basis), Premion®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Tetraammineplatinum (II) chloride hydrate
10836.03
10836.06
10836.03EA
9170
CZK
10836.03
10836.06
Tetraammineplatinum(II) chloride monohydrate ≥98% (metals basis), Premion®
Tetraammineplatinum(II) chloride monohydrate
Purity: 99.995%
CAS No.: 13933-33-0
Molecular Formula: Pt(NH3)4Cl2.H2O
Formula Weight: 352.13 (334.11anhy)
Physical Form: Crystalline
Density: 2.74
Sensitivity: Hygroscopic
Appearance: White to cream
Melting Point: 250°dec.
MDL No.: MFCD00149947
Vzorec:
Cl₂H₁₄N₄OPt MW: 352.123 g/mol Bod tání: 250 °C. (rozkl.) Hustota: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00149947 CAS číslo: 13933-33-0 EINECS: 237-706-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...