Tisk…

Potassium hexanitroiridate(III) (≥32,3% Ir)

Dodavatel: Thermo Fisher Scientific
Varování

10528.03 10528.06
10528.03EA 9240 CZK
10528.03 10528.06
Potassium hexanitroiridate(III) (≥32,3% Ir)
Potassium hexanitroiridate(III)

CAS No.: 38930-18-6 Molecular Formula: K3Ir(NO2)6 Formula Weight: 585.54 Physical Form: Micro Crystals MDL No.: MFCD00049672

Vzorec: K₃Ir(NO₂)₆
MW: 585.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049672
CAS číslo: 38930-18-6
EINECS: 254-191-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR