Tisk…

Dichloro(ethylenediamine)palladium(II) (44,8% Pd)

Dodavatel: Thermo Scientific
Varování

Synonyms: Dichloro(1,2-diaminoethane)palladium

10496.03 10496.06
10496.03EA 5980 CZK
10496.03 10496.06
Dichloro(ethylenediamine)palladium(II) (44,8% Pd)
Dichloro(ethylenediamine)palladium(II)

CAS No.: 15020-99-2 Molecular Formula: PdCl2(H2NCH2CH2NH2) Formula Weight: 237.41 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00044991

Vzorec: PdCl₂(H₂NCH₂CH₂NH₂)
MW: 237,41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044991
CAS číslo: 15020-99-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR