cis-Dichlorobis(diethylsulphide)platinum(II) (43,7% Pt)
Dodavatel: Thermo Scientific
|
Varování
|
10482.02
10482.03
10482.02EA
2850
CZK
10482.02
10482.03
cis-Dichlorobis(diethylsulphide)platinum(II) (43,7% Pt)
cis-Dichlorobis(diethylsulphide)platinum(II)
CAS No.: 15442-57-6
Molecular Formula: Pt[(C2H5)2S]2Cl2
Formula Weight: 446.37
Physical Form: Crystalline
Solubility: Soluble in acetone, alcohol and benzene
MDL No.: MFCD00050943
Applications: Hydrogenation
Vzorec:
Pt[(C₂H₅)₂S]₂Cl₂ MW: 446,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050943 CAS číslo: 15442-57-6 EINECS: 239-454-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...