Allylpalladium(II) chloride dimer (≥ 56,0% Pd)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Di-mu-chlorobis[(eta-allyl)palladium(II)], Allylpalladium(II) chloride, Allylpalladium chloride dimer
10005.01
10005.03
10005.06
10005.01EA
2020
CZK
10005.01
10005.03
10005.06
Allylpalladium(II) chloride dimer (≥ 56,0% Pd)
Allylpalladium(II) chloride dimer
CAS No.: 12012-95-2
Molecular Formula: C6H10Cl2Pd2
Formula Weight: 365.85
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow
Melting Point: 120° dec.
Solubility: Very soluble in dichloromethane and dichloroethane. Slightly soluble in toluene, insoluble in water
Beilstein Reference: 4124623
MDL No.: MFCD00044874
Vzorec:
C₆H₁₀Cl₂Pd₂ MW: 365,89 g/mol Bod tání: 120 °C (rozkl.) |
MDL Number:
MFCD00044874 CAS číslo: 12012-95-2 EINECS: 234-579-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...