Tisk…
Aspirator bottles
Dodavatel: Kavalier
632415046500 632415046940 632415046950 632415046956 632415046966 632415046967 632415046968
VTRB632415046500EA 0 CZK
VTRB632415046500 VTRB632415046940 VTRB632415046950 VTRB632415046956 VTRB632415046966 VTRB632415046967 VTRB632415046968
Aspirator bottles
Láhve Aspirátory
Glass, clear.
  • Interchangeable drain cock
  • Good chemical resistance
  • Good temperature resistance
With ground tube connector and ground drain cock. Neck with ground sleeve and ground glass stopper.