Tisk…
Safety shoes, lace-up, Style & Job, Tudor
Dodavatel: U-Power
BC20315/35 BC20315/36 BC20315/37 BC20315/38 BC20315/39 BC20315/40 BC20315/41 BC20315/42 BC20315/43 BC20315/44 BC20315/45 BC20315/46 BC20315/47 BC20315/48
114-3264EA 1770 CZK
114-3264 114-3265 114-3266 114-3267 114-3268 114-3269 114-3270 114-3271 114-3272 114-3273 114-3274 114-3275 114-3276 114-3277
Safety shoes, lace-up, Style & Job, Tudor
Obuv
Anti-static safety safety shoes made from soft suede leather with textile inserts and a slip-resistant and hard wearing PU sole.
  • Removable anatomical FlatFit insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Anthracite
Certifikace: EN 20345:2011; S1P, SRC