3,4,9,10-Perylenetetracarboxylic dianhydride for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Anthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone, Pigment Red 224, PTCDA
M-Clarity™ quality level = MQ200
8.20970.0025
8.20970.0100
8.20970.0025EA
1180
CZK
8.20970.0025
8.20970.0100
3,4,9,10-Perylenetetracarboxylic dianhydride for synthesis, Sigma-Aldrich®
3,4,9,10-Perylenetetracarboxylic dianhydride
Vzorec:
C₂₄H₈O₆ MW: 392,32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006916 CAS číslo: 128-69-8 EINECS: 204-905-3 |
Výsledky specifikačního testu
Solubility | 0,87 g/l (20 °C) |
Melting Point |
|
Molar Mass | 392.33 g/mol |
Bulk Density | 300 kg/m3 |
Flash Point |
|
pH | 6 - 7 (1 g/l, H2O, 20 °C) |
Bulk density | 300 kg/m3 |
Assay (acidimetric) | ≥98% |
Identity (IR) | Passes test |
Storage temperature | +15...+25 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...