Tisk…
Vacuum aspiration systems, AE and AK
Dodavatel: HAEP LABOR
800014900 800014800 800015400 800015500
HAEP800014900EA 23600 CZK
HAEP800014900 HAEP800014800 HAEP800015400 HAEP800015500
Vacuum aspiration systems, AE and AK
Čerpadla Vakuové aspirační systémy
The simple vacuum safety aspiration system, together with the quick lock adapters, filter, ventilation insert and the silicone tubing forms a complete unit. The comfortable aspiration devices additionally have a bottle holder and handle.
  • System optionally available with 2 or 4-litre bottle
  • Screw cap for the bottles with two quick action couplings that close automatically if the hose connections are disengaged
  • Extra-long (3 m) silicone tubing allows the bottle and the vacuum pump to be placed away from the workbench