Tisk…
VWR®, Test Sieves, 250×55 mm
Dodavatel: VWR Collection
510-3739EA 5110 CZK
510-3739 510-3740 510-3741 510-3742 510-3743 510-3744 510-3745 510-3746 510-3747 510-3748 510-3749 510-3750 510-3751 510-3752 510-3753 510-3754 510-3755 510-3756 510-3757 510-3758 510-3759 510-3760 510-3761 510-3762 510-3763 510-3764 510-3765 510-3766 510-3767 510-3768 510-3769 510-3770 510-3771 510-3772 510-3773 510-3774 510-3775 510-3776 510-3777 510-3778 510-3779 510-3780 510-3781 510-3782 510-3783 510-3784 510-3785 510-3786 510-3787 510-3788 510-3789 510-3790 510-3791 510-3792 510-3793 510-3794 510-3795 510-3796 510-3797 510-3798 510-3799 510-3800 510-3801 510-3802 510-3803 510-3804 510-3805 510-3806 510-3807 510-3808 510-3809 510-3810 510-3811 510-3812 510-3813 510-3715 510-3716 510-3717 510-3718 510-3719 510-3720 510-3721 510-3722 510-3723 510-3724 510-3725 510-3726 510-3727 510-3728 510-3729 510-3730 510-3731 510-3732 510-3733 510-3734 510-3735 510-3736 510-3737 510-3738
VWR®, Test Sieves, 250×55 mm
Síta Síta
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.