Tisk…
VWR®, Test Sieves, 400×65 mm, with Square Holes
Dodavatel: VWR Collection
510-3570EA 10760 CZK
510-3570 510-3571 510-3572 510-3573 510-3574 510-3575 510-3576 510-3577 510-3578 510-3579 510-3580 510-3581 510-3582 510-3583 510-3584 510-3585 510-3586 510-3587 510-3588 510-3589 510-3590 510-3591 510-3592 510-3593 510-3594 510-3595 510-3596 510-3597 510-3598 510-3599 510-3600 510-3601 510-3602 510-3603 510-3604 510-3605 510-3606 510-3607 510-3608 510-3609
VWR®, Test Sieves, 400×65 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.