Tisk…
Test sieves, 203×25 mm
Dodavatel: Retsch GmbH
60140000020 60.140.000025 60.140.000032 60.140.000036 60140000038 60.140.000040 60.140.000045 60.140.000050 60.140.000053 60140000056 60.140.000063 60.140.000071 60140000075 60.140.000080 60.140.000090 60140000106 60.140.000112 60140000125 60.140.000140 60.140.000150 60140000160 60.140.000180 60.140.000200 60140000212 60.140.000224 60140000250 60.140.000280 60140000300 60.140.000315 60.140.000355 60140000400 60140000425 60140000450 60140000500 60.140.000560 60.140.000630 60.140.000710 60.140.000800 60.140.000850 60.140.000900 60.140.001120 60.140.001180 60.140.001250 60140001400 60.140.001600 60140001700 60.140.001800 60.140.002000 60.140.002240 60140002360 60.140.002500 60.140.002800 60.140.003150 60.140.003350 60.140.003550 60140004000 60140004500 60.140.004750 60140005000 60.140.005600 60140006300 60.140.006700 60.140.007100 60.140.008000 60140009000 60.140.009500 60.140.010000 60140011200 60.140.012500 60.140.013200 60.140.014000 60.140.016000 60.140.018000 60.140.019000 60140020000 60140022400 60.140.025000 60140026500 60140028000 60.140.031500 60.140.035500 60140037500 60140040000 60.140.045000 60140050000 60.140.053000 60.140.056000 60.140.063000 60.140.071000 60.140.075000 60.140.080000 60.140.090000 60.140.100000 60.140.106000 60.140.112000 60140125000 60.140.001000 60.140.000100 60.140.000600 60.140.002360 60.140.000300 60.140.000020 60.140.000125 60.140.000500
510-1952EA 29520 CZK
510-1952 510-1857 510-1858 510-1859 510-1953 510-1860 510-1861 510-1862 510-1863 510-1958 510-1864 510-1865 510-1959 510-1866 510-1867 510-1960 510-1869 510-1961 510-1870 510-1871 510-1962 510-1872 510-1873 510-1963 510-1874 510-1964 510-1875 510-1965 510-1876 510-1877 510-1966 510-1967 510-1968 510-1969 510-1878 510-1879 510-1880 510-1881 510-1882 510-1883 510-1885 510-1886 510-1887 510-1970 510-1888 510-1971 510-1889 510-1890 510-1891 510-1972 510-1892 510-1893 510-1894 510-1895 510-1896 510-1973 510-1974 510-1897 510-1975 510-1898 510-1976 510-1899 510-1900 510-1909 510-1977 510-1910 510-1911 510-1978 510-1920 510-1921 510-1922 510-1923 510-1930 510-1931 510-1979 510-1980 510-1932 510-1986 510-1987 510-1933 510-1934 510-1988 510-1989 510-1935 510-1990 510-1936 510-1937 510-1938 510-1939 510-1940 510-1943 510-1944 510-1945 510-1946 510-1947 510-1992 510-1884 510-1868 RETS60.140.000600 RETS60.140.002360 RETS60.140.000300 RETS60.140.000020 RETS60.140.000125 RETS60.140.000500
Test sieves, 203×25 mm
Síta Síta
304 stainless steel, ISO 3310-1:2001-09.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieves are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.