Tisk…
ESD safety shoes, lace-up, Dimension Pro, Horizon Low
Dodavatel: ELTEN
727821/36 727821/37 727821/38 727821/39 727821/40 727821/41 727821/42 727821/43 727821/44 727821/45 727821/46 727821/47 727821/48
114-7718EA 2790 CZK
114-7718 114-7719 114-7720 114-7721 114-7722 114-7723 114-7724 114-7725 114-7726 114-7727 114-7728 114-7729 114-7730
ESD safety shoes, lace-up, Dimension Pro, Horizon Low
Obuv
Leather-free, anti-slip ESD safety shoes made from hydrophobised microfibre and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent) upper, a full padded bellows tongue plus a slip resistant, dual-density TPU/PU sole (Dimension Pro).
  • Climate-regulating, breathable textile lining
  • Semi-orthopaedic, anatomically-shaped insole supports the natural heel-to-toe foot movement
  • Replaceable insole; orthopaedic insoles possible
  • Soft fleece ESD insole
  • Sole prevents twisting and slipping
  • Metal-free, puncture resistant midsole
  • Protective steel
More Product Information