Tisk…

Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody (In Vivo Ready™) [clone: BNI3]

Dodavatel: Tonbo Biosciences
40-1529-U100 40-1529-U500
40-1529-U100EA 2380 CZK
40-1529-U100 40-1529-U500
Anti-CD152 (CTLA-4) Mouse Monoclonal Antibody (In Vivo Ready™) [clone: BNI3]
Protilátky
The BNI3 antibody is specific for human CD152, commonly known as CTLA-4, a 33 to 37 kDa protein expressed as a homodimer on the surface of activated T and B cells, and on thymocytes. CTLA-4 is structurally similar, yet functionally disparate, to the T cell co-stimulatory molecule CD28. Both CTLA-4 and CD28 interact with the co-stimulatory molecules CD80 (B7-1) and CD86 (B7-2) on antigen-presenting cells, with CTLA-4 displaying a higher avidity than CD28. While CD28 typically delivers a potent co-stimulatory signal in support of T cell activation, CTLA-4 appears to act as a negative regulator of T cell activation and may contribute to the suppressor function of Treg cells.

CTLA-4 proteins may be initially sequestered within Golgi vesicles, from which they can be transferred to and from the cell surface, a mechanism by which Treg cells can selectively impart suppressive functions. The BNI3 antibody may be used for flow cytometric analysis of intracellular or surface CTLA-4 expression, and is also widely used for neutralization of CTLA-4 when expressed at the cell surface. The BNI3 antibody is reported to be cross-reactive with Baboon, Cynomolgus and Rhesus CTLA-4. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD152 (CTLA-4)
Clonality: Monoclonal
Clone: BNI3
Conjugation: In Vivo Ready™
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Baboon, Cynomolgus, Rhesus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR