Tisk…

Anti-CD45 Mouse Antibody (redFluor™ 710) [clone: HI30]

Dodavatel: Tonbo Biosciences
80-0459-T025 80-0459-T100
80-0459-T025EA 751 CZK
80-0459-T025 80-0459-T100
Anti-CD45 Mouse Antibody (redFluor™ 710) [clone: HI30]
Protilátky
The HI30 antibody reacts with human CD45, one of the most abundant hematopoietic markers and one that is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.

The HI30 antibody is widely used as a marker for human CD45 expression on T cells, B cells, monocytes, macrophages, and NK cells.

Type: Primary
Antigen: CD45
Clonality:
Clone: HI30
Conjugation: redFluor™ 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR