Tisk…

Anti-Vtg Mouse Monoclonal Antibody [clone: BN-5]

Dodavatel: Cayman Chemical
170145-100 170145-500
CAYM170145-100EA 16920 CZK
CAYM170145-100 CAYM170145-500
Anti-Vtg Mouse Monoclonal Antibody [clone: BN-5]
Protilátky
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: BN-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Arctic Char, Atlantic Salmon, brown Trout, Flounder, Halibut, Rainbow Trout, Turbot, Wrasse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR